Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029857.3 | NP_084133.1 | 7, 8 | 1142 | ||
GenBank mRNA | AK014571.1 | - | 6, 7 | 743 | ||
AK029092.1 | - | 7, 8 | 1142 | |||
AK054337.1 | - | 7, 8 | 1139 | |||
AK170619.1 | - | 7, 8 | 871 | |||
AK171189.1 | - | 7, 8 | 888 | |||
BC013471.1 | - | 6, 7 | 768 | 4221072 |