Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9230110F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029863.1 | NP_084139.1 | 3 | 398 | ||
GenBank mRNA | AK020329.1 | - | 3 | 399 |
Gene Symbol: Gm5916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167587.1 | NP_001161059.1 | 3 | 405 |