Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 12, 13 | 1174 | ||
GenBank mRNA | AK122507.1 | - | 9, 10 | 2442 | ||
AK154554.1 | - | 12, 13 | 1170 | |||
AK208144.1 | - | 12 | ||||
BE862876.1 | - | 4, 5 | 347 |
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039520.1 | - | 4, 5 | 574 | ||
AK137412.1 | - | 5, 6 | 1948 | |||
BC032285.1 | - | 4, 5 | 1829 | |||
BC147312.1 | - | 1154 | 9056375 | |||
BC147313.1 | - | 1154 | 9056366 |
Gene Symbol: Tmem181c-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045744.1 | - | 4, 5 | 837 |