Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029868.2 | NP_084144.2 | 10, 11 | 2343 | ||
GenBank mRNA | AK037224.1 | - | 10, 11 | 2364 | ||
AK077148.1 | - | 10, 11 | 2313 | |||
AK133761.1 | - | 10, 11 | 2359 | |||
BC031428.1 | - | 9, 10 | 1416 | 4021073 | ||
BC066034.1 | - | 10, 11 | 2343 | 6836605 |