Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252625.1 | NP_001239554.1 | 4 | 572 | ||
NM_001252626.1 | NP_001239555.1 | 5 | 644 | |||
NM_029880.3 | NP_084156.2 | 4 | 515 | |||
GenBank mRNA | AK007522.1 | - | 5 | 527 | ||
AK020666.1 | - | 4 | 444 | |||
AK021033.1 | - | 4 | 407 | |||
AK036168.1 | - | 4 | 437 | |||
AK040396.1 | - | 5 | 576 | |||
AK145232.1 | - | 4 | 463 | |||
AK159187.1 | - | 4 | 411 | |||
AK159932.1 | - | 4 | 412 | |||
AK168895.1 | - | 4 | 406 | |||
BC021466.1 | - | 4 | 461 | 5068419 |