Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029896.1 | NP_084172.1 | 5, 6 | 843 | ||
GenBank mRNA | AK035109.1 | - | 5, 6 | 845 | ||
AK037620.1 | - | 5, 6 | 850 | |||
AK149692.1 | - | 5, 6 | 844 | |||
AK159240.1 | - | 5, 6 | 843 | |||
BC019115.1 | - | 4, 5 | 369 | |||
BC031502.1 | - | 5, 6 | 533 | |||
BC094419.1 | - | 5, 6 | 806 | |||
BC157943.1 | - | 766 | 9007278 | |||
BC157945.1 | - | 767 | 9007280 | |||
BC158054.1 | - | 767 | 9007356 | |||
EF011615.1 | - | 5, 6 | 532 |