Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 3 | 691 | ||
NM_001168404.1 | NP_001161876.1 | 3 | 228 | |||
NM_001168405.1 | NP_001161877.1 | 2 | 585 | |||
NM_029898.3 | NP_084174.2 | 2 | 585 | |||
GenBank mRNA | AK039553.1 | - | 2 | 586 | ||
AK045480.1 | - | 2 | 219 | |||
AK081197.1 | - | 3 | 229 | |||
AK138273.1 | - | 2 | 240 | |||
AK140417.1 | - | 3 | 554 | |||
AK155092.1 | - | 1 | 26 | |||
AK171119.1 | - | 1 | 26 | |||
BC152317.1 | - | 2 | 212 | 40088531 | ||
BC152536.1 | - | 2 | 212 | 40088530 |