Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289999.1 | NP_001276928.1 | 2 | 602 | ||
NM_001290000.1 | NP_001276929.1 | 2 | 454 | |||
NM_005384.2 | NP_005375.2 | 2 | 459 | |||
XM_017014743.1 | XP_016870232.1 | 2 | 597 | |||
XM_017014744.1 | XP_016870233.1 | 2 | 366 | |||
GenBank mRNA | AK313970.1 | - | 2 | 276 | ||
BC008197.1 | - | 1 | 97 | 3048714 | ||
BQ937243.1 | - | 2 | 459 | |||
CR542049.1 | - | 1 | 64 | |||
HM204564.1 | - | 1 | 64 | |||
HY038941.1 | - | 2 | 429 | |||
S79880.1 | - | 2 | 277 | |||
U26173.1 | - | 2 | 277 | |||
X64318.1 | - | 2 | 277 |