Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082485.1 | NP_001075954.1 | 9 | 2210 | ||
NM_001135019.1 | NP_001128491.1 | 9 | 2199 |