Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082485.1 | NP_001075954.1 | 9 | 1025 | ||
NM_001135019.1 | NP_001128491.1 | 9 | 1014 | |||
GenBank mRNA | BC048701.1 | - | 9 | 965 | ||
BC061209.1 | - | 9 | 1011 | |||
BC089552.1 | - | 9 | 1024 |