Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001980.2 | NP_001001980.2 | 8 | 865 | ||
NM_001256122.1 | NP_001243051.1 | 3 | 627 | |||
GenBank mRNA | AK019457.1 | - | 3 | 484 | ||
AK140954.1 | - | 3 | 393 | |||
AK147547.1 | - | 3 | 628 | |||
CD804084.1 | - | 3 | 325 |