Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001980.2 | NP_001001980.2 | 25 | 3177 | ||
NM_001256122.1 | NP_001243051.1 | 20 | 2939 | |||
GenBank mRNA | AK122443.1 | - | 18 | 2292 | ||
AK134095.1 | - | 20 | 2740 | |||
AK147547.1 | - | 20 | 2940 | |||
BC042189.1 | - | 17 | 2256 |