Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029930.2 | NP_084206.1 | 7 | 2629 | ||
GenBank mRNA | AK004097.1 | - | 1 | 128 | ||
AK014379.1 | - | 7 | 2158 | |||
AK019001.1 | - | 1 | 94 | |||
AK044692.1 | - | 2 | 481 | |||
AK053509.1 | - | 7 | 2630 | |||
AK083917.1 | - | 7 | 2577 | |||
AK132148.1 | - | 7 | 2597 | |||
AK148593.1 | - | 7 | 2608 | |||
AK173013.1 | - | 7 | 2611 | |||
BC031718.1 | - | 7 | 2657 | 3156550 | ||
BC043304.1 | - | 5 | 889 |