Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029935.5 | NP_084211.2 | 6 | 1769 | ||
GenBank mRNA | AB187269.1 | - | 5 | 1209 | ||
AF320786.1 | - | 5 | 1260 | |||
AK019474.1 | - | 6 | 1724 | |||
AK031745.1 | - | 4 | 470 | |||
AK035637.1 | - | 6 | 1857 | |||
AK043803.1 | - | 4 | 617 | |||
AK082318.1 | - | 6 | 1835 | |||
AK083614.1 | - | 6 | 1769 | |||
AK122324.1 | - | 6 | 2070 | |||
BC031443.1 | - | 6 | 1726 | 4240286 |