Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029935.5 | NP_084211.2 | 2 | 1020 | ||
GenBank mRNA | AB187269.1 | - | 1 | 460 | ||
AF320786.1 | - | 1 | 511 | |||
AK019474.1 | - | 2 | 975 | |||
AK035637.1 | - | 2 | 1108 | |||
AK082318.1 | - | 2 | 1086 | |||
AK083614.1 | - | 2 | 1020 | |||
AK122324.1 | - | 2 | 1321 | |||
BC031443.1 | - | 2 | 977 | 4240286 |