Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077494.3 | NP_001070962.1 | 10 | 1059 | ||
NM_001261403.2 | NP_001248332.1 | 9 | 1154 | |||
NM_001288724.1 | NP_001275653.1 | 10 | 1353 | |||
NM_001322934.1 | NP_001309863.1 | 10 | 1063 | |||
NM_001322935.1 | NP_001309864.1 | 10 | 1353 | |||
NM_002502.5 | NP_002493.3 | 10 | 1063 | |||
XM_011539830.2 | XP_011538132.1 | 5 | 417 | |||
XM_011539831.2 | XP_011538133.1 | 5 | 414 | |||
XM_017016278.1 | XP_016871767.1 | 10 | 1348 | |||
GenBank mRNA | AK292654.1 | - | 9 | 1007 | ||
BC002844.2 | - | 10 | 890 | 3634575 | ||
BT009769.1 | - | 9 | 790 | |||
S76638.1 | - | 10 | 1040 | |||
U09609.2 | - | 9 | 790 | |||
X61498.1 | - | 10 | 953 | |||
X61499.1 | - | 10 | 953 |