Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029936.2 | NP_084212.2 | 6 | 814 | ||
GenBank mRNA | AK019495.1 | - | 6 | 794 | ||
AK045032.1 | - | 6 | 779 | |||
AK052540.1 | - | 6 | 794 | |||
BC023303.1 | - | 6 | 762 | |||
BC049261.1 | - | 6 | 755 | |||
BC055481.1 | - | 5 | 532 | |||
BC080729.1 | - | 6 | 782 | 6504816 |