Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198957.2 | NP_945195.1 | 4 | 2736 | ||
GenBank mRNA | AK080943.1 | - | 4 | 2666 | ||
AK169895.1 | - | 2 | 2160 | |||
BC030408.1 | - | 1 | 1073 | |||
BC080741.1 | - | 3 | 2651 | 30461213 | ||
BC137620.1 | - | 3 | 2751 | 8860639 |