Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029938.1 | NP_084214.1 | 3 | 220 | ||
GenBank mRNA | AK021267.1 | - | 3 | 143 | ||
AK155105.1 | - | 3 | 223 | |||
AK168110.1 | - | 3 | 192 | |||
AK178770.1 | - | 1 | 23 | |||
AK187484.1 | - | 2 | 67 | |||
BC147293.1 | - | 2 | 179 | 9056353 | ||
BC147294.1 | - | 3 | 205 | 9056344 | ||
BC147741.1 | - | 3 | 205 | 9056690 | ||
BC147751.1 | - | 3 | 205 | 9056700 | ||
CJ069840.1 | - | 3 | 221 |