Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramef12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029948.2 | NP_084224.1 | 3 | 592 | ||
GenBank mRNA | AK019795.1 | - | 3 | 593 | ||
AK167683.1 | - | 3 | 410 | |||
BC066168.1 | - | 3 | 547 | 30474135 |