Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198297.3 | NP_938039.2 | 2 | 111 | ||
GenBank mRNA | AJ297969.1 | - | 2 | 129 | ||
AK030807.1 | - | 2 | 294 | |||
AK138735.1 | - | 177 | ||||
AK163002.1 | - | 1 | 225 | |||
BC104350.1 | - | 2 | 111 | 40049557 | ||
BC104351.1 | - | 2 | 111 | 40049558 |