Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029953.1 | NP_084229.1 | 4 | 586 | ||
GenBank mRNA | BC049886.1 | - | 4 | 586 | 5387778 |