Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029958.1 | NP_084234.1 | 4 | 416 | ||
GenBank mRNA | AK020307.1 | - | 3 | 322 | ||
AY299493.1 | - | 4 | 416 | |||
BC111100.1 | - | 4 | 409 | 40049540 |