Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029961.2 | NP_084237.1 | 22 | 2748 | ||
GenBank mRNA | AK139832.1 | - | 9 | 1142 | ||
AY766238.1 | - | 2 | 245 | |||
AY766239.1 | - | 22 | 2712 | |||
JQ655148.1 | - | 9 | 2227 |