Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029965.2 | NP_084241.1 | 2 | 334 | ||
GenBank mRNA | AK020104.1 | - | 2 | 239 | ||
AK036051.1 | - | 2 | 193 | |||
AK042063.1 | - | 2 | 331 | |||
AK044579.1 | - | 340 | ||||
AK080591.1 | - | 2 | 134 | |||
AK152760.1 | - | 2 | 324 | |||
AK153658.1 | - | 2 | 335 | |||
AK162554.1 | - | 2 | 348 | |||
BC031383.1 | - | 2 | 148 | |||
BC114210.1 | - | 2 | 334 | 6309632 | ||
BC138930.1 | - | 2 | 162 | 8861952 | ||
BC145718.1 | - | 2 | 167 | 40131021 |