Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029975.2 | NP_084251.1 | 3 | 625 | ||
GenBank mRNA | AK020784.1 | - | 3 | 500 | ||
AK037408.1 | - | 3 | 597 | |||
AK084712.1 | - | 724 | ||||
AK085119.1 | - | 787 | ||||
AY249021.1 | - | 2 | 254 | |||
BC132070.1 | - | 3 | 625 | 40130347 | ||
BC132072.1 | - | 3 | 625 | 40130349 | ||
EU047812.1 | - | 3 | 489 |