Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029981.1 | NP_084257.1 | 11 | 1704 | ||
GenBank mRNA | AB096928.1 | - | 11 | 1704 | ||
AK156991.1 | - | 11 | 1707 | |||
BC128568.1 | - | 11 | 1656 | 40097415 | ||
BC129984.1 | - | 11 | 1656 | 40097418 |