Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133922.3 | NP_598683.3 | 6 | 968 | ||
GenBank mRNA | AB041543.1 | - | 5 | 798 | ||
AK040525.1 | - | 6 | 970 | |||
AK128931.1 | - | 6 | 919 | |||
AK162934.1 | - | 6 | 676 | |||
AK173281.1 | - | 6 | 2672 | |||
BC066816.1 | - | 6 | 888 | 30084911 |