Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 1528 | |||
GenBank mRNA | AK153696.1 | - | 2118 | |||
AK173208.1 | - | 2299 | ||||
BC070432.1 | - | 1528 | ||||
BC100371.1 | - | 1 | 682 |