Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133777.2 | NP_598538.1 | 4 | 597 | ||
GenBank mRNA | AK003078.1 | - | 4 | 611 | ||
AK017648.1 | - | 4 | 613 | |||
AK020143.1 | - | 610 | ||||
AK146276.1 | - | 4 | 609 | |||
AK154026.1 | - | 4 | 610 | |||
AK154067.1 | - | 4 | 613 | |||
BC012255.1 | - | 4 | 597 | 3581845 | ||
BC030171.1 | - | 4 | 591 | 5037250 | ||
BC083323.1 | - | 4 | 579 | 5715696 |