Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318758.1 | NP_001305687.1 | 2, 3 | 1195 | ||
NM_002504.5 | NP_002495.2 | 2, 3 | 1195 | |||
NM_147134.3 | NP_667345.1 | 2, 3 | 1195 | |||
GenBank mRNA | AF332009.1 | - | 2, 3 | 1100 | ||
AK225750.1 | - | 2, 3 | 1068 | |||
AK291643.1 | - | 2, 3 | 1085 | |||
AL832342.1 | - | 2, 3 | 1078 | |||
BC006447.1 | - | 1, 2 | 692 | |||
BC012151.1 | - | 2, 3 | 1065 | 4558442 | ||
U15306.1 | - | 1, 2 | 991 |