Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175268.4 | NP_780477.2 | 3 | 357 | ||
NM_212473.1 | NP_997638.1 | 3 | 420 | |||
GenBank mRNA | AK020830.1 | - | 3 | 358 | ||
AK031576.1 | - | 2 | 201 | |||
AK051299.1 | - | 3 | 421 | |||
AK147350.1 | - | 3 | 753 | |||
AK147725.1 | - | 3 | 549 | |||
AK156629.1 | - | 3 | 320 | |||
AK165022.1 | - | 414 | ||||
AK180944.1 | - | 2 | 174 | |||
AK187644.1 | - | 2 | 174 | |||
AK195672.1 | - | 2 | 125 | |||
AK207193.1 | - | 2 | 174 | |||
BC035523.1 | - | 3 | 352 | 5400742 |