Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030017.3 | NP_084293.1 | 4 | 528 | ||
GenBank mRNA | AK020927.1 | - | 4 | 525 | ||
AK039233.1 | - | 4 | 529 | |||
BC016204.1 | - | 4 | 475 | 4527851 | ||
BC067213.1 | - | 3 | 1649 |