Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081030.2 | NP_001074499.2 | 29 | 4177 | ||
NM_001170561.1 | NP_001164032.1 | 30 | 4255 | |||
GenBank mRNA | AK129485.1 | - | 29 | 4121 | ||
AK147332.1 | - | 29 | 4134 | |||
BC098209.1 | - | 10 | 1520 | |||
BC157935.1 | - | 29 | 4067 | 9088115 | ||
BC158013.1 | - | 30 | 4145 | 9007320 | ||
BC172094.1 | - | 29 | 4067 | 9054380 |