Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFYC SIRNA Assays ›
Gene Symbol:
NFYC
LOC494127
Gene Name:
nuclear transcription factor Y subunit gamma
NFYC pseudogene
Gene Aliases:
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
-
Chromosome Location:
Chr.1: 40691570 - 40771603 on Build GRCh38
Chromosome Location:
Chr.9: 87083886 - 87085225 on Build GRCh38
UniGene ID:
Hs.713051
Hs.626316
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s9535
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFYC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142587.1
NP_001136059.1
3
449
NM_001142588.1
NP_001136060.1
3
449
NM_001142589.1
NP_001136061.1
3
449
NM_001142590.1
NP_001136062.1
8
826
NM_001308114.1
NP_001295043.1
2
151
NM_001308115.1
NP_001295044.1
3
445
NM_014223.4
NP_055038.2
3
449
XM_005270894.1
XP_005270951.1
3
445
XM_005270895.3
XP_005270952.3
3
301
XM_006710658.2
XP_006710721.2
3
301
XM_006710660.2
XP_006710723.2
3
301
XM_006710661.1
XP_006710724.1
3
330
XM_006710662.2
XP_006710725.2
3
301
XM_011541516.1
XP_011539818.1
3
250
XM_011541517.1
XP_011539819.1
3
301
XM_017001364.1
XP_016856853.1
3
470
XM_017001365.1
XP_016856854.1
3
330
XM_017001366.1
XP_016856855.1
3
404
XM_017001367.1
XP_016856856.1
3
445
XM_017001368.1
XP_016856857.1
3
445
XM_017001369.1
XP_016856858.1
4
802
GenBank mRNA
AB208975.1
-
3
276
AF191744.1
-
2
150
AK000346.1
-
3
402
AK127677.1
-
8
821
AK300774.1
-
3
252
AK301385.1
-
3
333
BC005003.1
-
3
298
3502903
BP230898.1
-
3
445
BT020081.1
-
2
143
D85425.1
-
3
226
D89986.1
-
3
211
DC297902.1
-
5
561
DC391720.1
-
3
449
DC428152.1
-
3
449
U62296.1
-
2
143
U78774.1
-
4
317
Z74792.1
-
3
328
Gene Symbol: LOC494127
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_036691.1
1
217
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012866, XM_006238785, U17607, BC093619.
This siRNA also targets the following transcripts in Mouse: NM_008692, NM_001048168, NM_001277095, AB006181, BC085261, and others.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein folding
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-82rcf:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline