Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175271.4 | NP_780480.2 | 3 | 649 | ||
GenBank mRNA | AK045289.1 | - | 3 | 619 | ||
AK051709.1 | - | 3 | 620 | |||
BC052178.1 | - | 3 | 640 | |||
BC069178.1 | - | 3 | 561 | |||
BC138893.1 | - | 3 | 450 | 8861915 | ||
BC138894.1 | - | 3 | 450 | 8861916 |