Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030055.1 | NP_084331.1 | 1, 2 | 129 | ||
GenBank mRNA | AK019803.1 | - | 1, 2 | 130 |