Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030058.2 | NP_084334.1 | 2 | 307 | ||
GenBank mRNA | AK020315.1 | - | 2 | 280 | ||
AY226988.1 | - | 2 | 307 | |||
BC119299.1 | - | 2 | 277 | 8734048 | ||
BC119301.1 | - | 2 | 277 | 8734050 |