Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291126.1 | NP_001278055.1 | 4 | 335 | ||
NM_001291127.1 | NP_001278056.1 | 3 | 351 | |||
NM_030064.4 | NP_084340.2 | 4 | 504 | |||
GenBank mRNA | AK019135.1 | - | 4 | 492 | ||
AK029376.1 | - | 4 | 285 | |||
AK031159.1 | - | 4 | 253 | |||
AK170022.1 | - | 3 | 352 | |||
AK170729.1 | - | 4 | 421 | |||
AY099326.1 | - | 4 | 250 | |||
BM947867.1 | - | 3 | 274 | |||
BY262829.1 | - | 4 | 312 | |||
CA750740.1 | - | 4 | 484 | |||
DV662694.1 | - | 4 | 463 |