Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291125.1 | NP_001278054.1 | 4 | 512 | ||
NM_001291126.1 | NP_001278055.1 | 4 | 544 | |||
NM_001291127.1 | NP_001278056.1 | 3 | 560 | |||
NM_030064.4 | NP_084340.2 | 4 | 713 | |||
GenBank mRNA | AK019135.1 | - | 4 | 701 | ||
AK029376.1 | - | 4 | 494 | |||
AK031159.1 | - | 4 | 462 | |||
AK170022.1 | - | 3 | 561 | |||
AK170729.1 | - | 4 | 630 | |||
AY099326.1 | - | 4 | 459 | |||
AY099327.1 | - | 4 | 258 | |||
BC033533.1 | - | 4 | 402 | 2655211 | ||
BM947867.1 | - | 3 | 483 | |||
DV662694.1 | - | 4 | 672 |