Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166377.1 | NP_001159849.1 | 6 | 1569 | ||
NM_001166378.1 | NP_001159850.1 | 4 | 1268 | |||
NM_001166379.1 | NP_001159851.1 | 4 | 1384 | |||
NM_001166380.1 | NP_001159852.1 | 5 | 1360 | |||
NM_030066.3 | NP_084342.1 | 6 | 1453 | |||
GenBank mRNA | AK019405.1 | - | 6 | 1380 | ||
AK147002.1 | - | 6 | 1520 | |||
BC021410.1 | - | 5 | 1288 | 5007058 | ||
BC026488.1 | - | 4 | 1352 | 4504219 | ||
BC068228.1 | - | 6 | 1402 | 6413546 |