Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 9 | 925 | ||
NM_001159966.1 | NP_001153438.1 | 9 | 823 | |||
NM_001159967.1 | NP_001153439.1 | 9 | 925 | |||
NM_001159968.1 | NP_001153440.1 | 8 | 820 | |||
NM_023884.4 | NP_076373.3 | 9 | 865 | |||
GenBank mRNA | AF312924.1 | - | 8 | 773 | ||
AK018622.1 | - | 9 | 867 | |||
AK019543.1 | - | 8 | 823 | |||
AK029049.1 | - | 9 | 866 | |||
AK029993.1 | - | 10 | 1003 | |||
AK033549.1 | - | 9 | 821 | |||
AK036803.1 | - | 8 | 797 | |||
AK161080.1 | - | 9 | 892 | |||
AK165002.1 | - | 9 | 817 | |||
AK169368.1 | - | 9 | 843 | |||
BC043132.1 | - | 9 | 830 | 6811441 | ||
BC052663.1 | - | 9 | 842 | 30051209 |