Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153502.3 | NP_705722.2 | 3 | 226 | ||
GenBank mRNA | AF492400.1 | - | 3 | 226 | ||
BC022973.1 | - | 385 | ||||
BC023221.1 | - | 3 | 203 | |||
BC132370.1 | - | 3 | 236 | 40130647 | ||
BC138797.1 | - | 3 | 236 | 8861819 | ||
BC145421.1 | - | 3 | 236 | 9053979 |