Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083891.1 | NP_001077360.1 | 3 | 289 | ||
NM_030087.2 | NP_084363.2 | 4 | 1381 | |||
GenBank mRNA | AK077433.1 | - | 3 | 230 | ||
AK154191.1 | - | 4 | 1314 | |||
AK154477.1 | - | 4 | 1360 | |||
AK201972.1 | - | 1 | 33 | |||
BC013640.1 | - | 4 | 1341 | 4188234 | ||
BU555486.1 | - | 3 | 292 |