Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030093.3 | NP_084369.2 | 3, 4 | 443 | ||
GenBank mRNA | AK019452.1 | - | 3, 4 | 444 | ||
BC028766.1 | - | 3, 4 | 384 | 1244556 |