Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024137.1 | NP_001019308.1 | 6 | 513 | ||
GenBank mRNA | AB195276.1 | - | 6 | 513 | ||
AK145985.1 | - | 6 | 514 | |||
AK167492.1 | - | 6 | 481 | |||
BC113160.1 | - | 6 | 510 | 40060148 | ||
BC113770.1 | - | 6 | 510 | 40060146 |