Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033186.3 | NP_001028358.1 | 4 | 317 | ||
NM_001177898.1 | NP_001171369.1 | 4 | 317 | |||
NM_001177899.1 | NP_001171370.1 | 4 | 317 | |||
GenBank mRNA | AK040068.1 | - | 4 | 317 | ||
AK041917.1 | - | 4 | 280 | |||
AK137792.1 | - | 4 | 285 | |||
AK137942.1 | - | 4 | 285 | |||
AK138030.1 | - | 4 | 300 | |||
AK169685.1 | - | 4 | 290 | |||
AK169825.1 | - | 4 | 282 | |||
BC145402.1 | - | 4 | 246 | 9053960 | ||
BC150764.1 | - | 4 | 246 | 9087675 | ||
BC171961.1 | - | 4 | 246 | 9054247 | ||
BY723539.1 | - | 4 | 297 |