Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030110.1 | NP_084386.1 | 12 | 1336 | ||
GenBank mRNA | BC125419.1 | - | 11 | 930 | ||
BC125445.1 | - | 11 | 930 | |||
BC145173.1 | - | 10 | 891 |