Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002508.2 | NP_002499.2 | 16 | 3225 | ||
XM_011544195.2 | XP_011542497.1 | 15 | 3096 | |||
GenBank mRNA | AB209448.1 | - | 16 | 3107 | ||
BC045606.1 | - | 13 | 2736 | 5271590 | ||
CN267119.1 | - | 2 | 175 | |||
M27445.1 | - | 8 | 1125 | |||
M30269.1 | - | 16 | 3213 |