Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170710.1 | NP_001164181.1 | 4 | 794 | ||
NM_001170711.1 | NP_001164182.1 | 4 | 800 | |||
NM_030121.4 | NP_084397.3 | 4 | 790 | |||
GenBank mRNA | AF398969.1 | - | 4 | 717 | ||
AK016001.1 | - | 4 | 848 | |||
AK021239.1 | - | 4 | 733 | |||
AK030168.1 | - | 4 | 733 | |||
BC025106.1 | - | 4 | 714 | 4194829 |